p-Toluenesulfonyl chloride 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
4-Toluenesulfonyl chloride, Toluene-p-sulfonyl chloride, Toluene-4-sulphonyl chloride, Tosyl chloride, 4-Toluenesulphonyl chloride, Toluene-4-sulfonyl chloride, Toluene-p-sulphonyl chloride
MDL: MFCD00007450
Beilstein: 607898
Soluble in alcohol, benzene, and ether. Insoluble in water
Purity: 98%
CAS No.: 98-59-9
Molecular Formula: C7H7ClO2S
Formula Weight: 190.65
Flash Point: 128°(262°F)
Melting Point: 67-70°
Boiling Point: 146°/15mm
Solubility: Soluble in alcohol, benzene, and ether. Insoluble in water
Beilstein Reference: 607898
Merck Reference: 14,9534
UN No.: 3261
MDL No.: MFCD00007450
Formula:
C₇H₇ClO₂S MW: 190.65 g/mol Boiling Pt: 265 Melting Pt: 67 Density: 1.35 Flash Pt: 128 °C (262 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007450 CAS Number: 98-59-9 EINECS: 202-684-8 UN: 3261 ADR: 8 III Merck Index: 14,9534 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...