2-Oxazolidinone 99%

Supplier: Thermo Scientific Chemicals

Synonyms: 2-Oxazolidone, Oxazolidin-2-one

A14543-36 A14543-14 A14543-22
AAA14543-36EA 446 USD
AAA14543-36 AAA14543-14 AAA14543-22
2-Oxazolidinone 99%
2-Oxazolidinone

Fieser: 12,359


Purity: 99% CAS No.: 497-25-6 Molecular Formula: C3H5NO2 Formula Weight: 87.08 Melting Point: 84-89° Boiling Point: 220°/48mm MDL No.: MFCD00005268

Formula: C₃H₅NO₂
MW: 87.08 g/mol
Boiling Pt: 220 °C (48 mmHg)
Melting Pt: 84…89 °C
Storage Temperature: Ambient
MDL Number: MFCD00005268
CAS Number: 497-25-6
EINECS: 207-840-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR