(±)-3-Octanol 97%

Supplier: Thermo Scientific Chemicals
Warning

A14488-30 A14488-18
AAA14488-30EA 162 USD
AAA14488-30 AAA14488-18
(±)-3-Octanol 97%
(±)-3-Octanol

Purity: 97% CAS No.: 589-98-0 Molecular Formula: C8H18O Formula Weight: 130.23 Flash Point: 68°(154°F) Refractive Index: 1.4270 Odor: Pleasant Melting Point: -45° Boiling Point: 175-176° Beilstein Reference: 1719310 MDL No.: MFCD00004590

Formula: C₈H₁₈O
Boiling Pt: 175…176 °C
Melting Pt: –45 °C
Density: 0.820
Flash Pt: 68 °C(154 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004590
CAS Number: 589-98-0
EINECS: 209-667-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR