Indole 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1H-Indole

A14427-0B A14427-18 A14427-30
AAA14427-0BEA 525 USD
AAA14427-0B AAA14427-18 AAA14427-30
Indole 99%
Indole

Purity: 99% CAS No.: 120-72-9 Molecular Formula: C8H7N Formula Weight: 117.15 Flash Point: 121°(249°F) Odor: Unpleasant pH: 6 Melting Point: 50-54° Boiling Point: 253-254° Beilstein Reference: 107693 Merck Reference: 14,4963 UN No.: 2811 MDL No.: MFCD00005607

Formula: C₈H₇N
MW: 117.15 g/mol
Boiling Pt: 253…254 °C
Melting Pt: 50…54 °C
Density: 1.220
Flash Pt: 121 °C (249 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005607
CAS Number: 120-72-9
EINECS: 204-420-7
UN: 2811
ADR: 6.1 III
Merck Index: 14,4963

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR