(±)-Mandelic acid 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
DL-Amygdalic Acid, Mandelic acid, DL-Mandelic acid, 2-Hydroxy-2-phenylacetic acid, (±)-2-Hydroxy-2-phenylacetic acid, DL-2-Hydroxy-2-phenylacetic acid
A14223-36
A14223-0E
A14223-22
AAA14223-36EA
107
USD
AAA14223-36
AAA14223-0E
AAA14223-22
(±)-Mandelic acid 99%
(±)-Mandelic acid
Purity: 99%
CAS No.: 90-64-2
Molecular Formula: C8H8O3
Formula Weight: 152.15
Physical Form: Cryst powder
Sensitivity: Light Sensitive
Appearance: White
Melting Point: 118-121°
Beilstein Reference: 510011
Merck Reference: 14,5717
MDL No.: MFCD00064250
Formula:
C₈H₈O₃ Boiling Pt: 322 °C (1013 hPa) Melting Pt: 118…121 °C Density: 1.361 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00064250 CAS Number: 90-64-2 EINECS: 202-007-6 Merck Index: 14,5717 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...