2-Mercaptopyridine-N-oxide 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Mercaptopyridine 1-oxide

A14152-14 A14152-06 A14152-22
AAA14152-14EA 211 USD
AAA14152-14 AAA14152-06 AAA14152-22
2-Mercaptopyridine-N-oxide 99%
2-Mercaptopyridine N-oxide

Fieser: 12,417 14,268 18,187 19,170


Purity: 99% CAS No.: 1121-31-9 Molecular Formula: C5H5NOS Formula Weight: 127.17 Melting Point: 69-72° Beilstein Reference: 906983 Merck Reference: 14,7994 MDL No.: MFCD00006196

Formula: C₅H₅NOS
Melting Pt: 69…72 °C
Storage Temperature: Ambient
MDL Number: MFCD00006196
CAS Number: 1121-31-9
EINECS: 214-329-4
Merck Index: 14,7994

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR