2-Mercaptopyridine-N-oxide 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Mercaptopyridine 1-oxide
A14152-14
A14152-06
A14152-22
AAA14152-14EA
211
USD
AAA14152-14
AAA14152-06
AAA14152-22
2-Mercaptopyridine-N-oxide 99%
2-Mercaptopyridine N-oxide
Fieser: 12,417 14,268 18,187 19,170
Purity: 99%
CAS No.: 1121-31-9
Molecular Formula: C5H5NOS
Formula Weight: 127.17
Melting Point: 69-72°
Beilstein Reference: 906983
Merck Reference: 14,7994
MDL No.: MFCD00006196
Formula:
C₅H₅NOS Melting Pt: 69…72 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006196 CAS Number: 1121-31-9 EINECS: 214-329-4 Merck Index: 14,7994 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...