Phosphinic acid (Hypophosphorous acid) 50% (w/w) in aqueous solution

Supplier: Thermo Scientific Chemicals
Danger

014142-18 014142-36
AAA14142-18EA 76.65 USD
AAA14142-18 AAA14142-36
Phosphinic acid (Hypophosphorous acid) 50% (w/w) in aqueous solution
Phosphinic acid (Hypophosphorous acid)

Liquid


CAS No.: 6303-21-5 Molecular Formula: H3O2P Formula Weight: 66.00 Physical Form: Liquid Density: 1.225 Appearance: Colorless Odor: Pungent UN No.: 3264 MDL No.: MFCD02183592

Formula: H₃O₂P
MW: 66 g/mol
Density: 1.225
Storage Temperature: Ambient
MDL Number: MFCD02183592
CAS Number: 6303-21-5
EINECS: 228-601-5
UN: 3264
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR