4-Aminobenzyl alcohol 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-(Hydroxymethyl)aniline, (4-Aminophenyl)methanol
A14130-14
A14130-06
AAA14130-14EA
446
USD
AAA14130-14
AAA14130-06
4-Aminobenzyl alcohol 98%
4-Aminobenzyl alcohol
Purity: 98%
CAS No.: 623-04-1
Molecular Formula: C7H9NO
Formula Weight: 123.16
Storage Temperature: 2°C to 8°C
Melting Point: 59-63°
Beilstein Reference: 2078680
MDL No.: MFCD00014782
Formula:
C₇H₉NO MW: 123.15 Boiling Pt: 169…171 °C (11 mmHg) Melting Pt: 75 Storage Temperature: Refrigerator |
MDL Number:
MFCD00014782 CAS Number: 623-04-1 EINECS: 210-767-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...