Benzoyl chloride 99+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Benzoic acid chloride
A14107-0B
A14107-30
A14107-0E
AAA14107-0BEA
75.94
USD
AAA14107-0B
AAA14107-30
AAA14107-0E
Benzoyl chloride 99+%
Benzoyl chloride
Purity: 99+%
CAS No.: 98-88-4
Molecular Formula: C7H5ClO
Formula Weight: 140.57
Flash Point: 69°(156°F)
Physical Form: Liquid
Density: 1.211
Refractive Index: 1.5535
Sensitivity: Moisture Sensitive
Appearance: Colorless
Odor: Pungent
pH: 2
Melting Point: -1°
Boiling Point: 197-199°
Merck Reference: 14,1112
UN No.: 1736
MDL No.: MFCD00000653
Formula:
C₇H₅ClO Boiling Pt: 197…199 °C Melting Pt: –1 °C Density: 1.211 Flash Pt: 69 °C(156 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000653 CAS Number: 98-88-4 EINECS: 202-710-8 UN: 1736 ADR: "8",II Merck Index: 14,1112 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...