2,4-Difluorophenol 98+%

Supplier: Thermo Scientific Chemicals
Danger

A14044-30 A14044-18
AAAA14044-30EA 469 USD
AAAA14044-30 AAA14044-18
2,4-Difluorophenol 98+%
2,4-Difluorophenol
Formula: C₆H₄F₂O
Boiling Pt: 52…54 °C/14 mm
Melting Pt: 21…24 °C
Density: 1.362
Flash Pt: 56 °C (132 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009715
CAS Number: 367-27-1
EINECS: 206-688-0
UN: 2925
ADR: 4.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR