4-Aminoantipyrine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Amino-2,3-dimethyl-1-phenyl-3-pyrazolin-5-one, Ampyrone, 4-Aminophenazone
A13846-36
A13846-0E
A13846-22
AAA13846-36EA
241
USD
AAA13846-36
AAAA13846-0E
AAA13846-22
4-Aminoantipyrine 97%
4-Aminoantipyrine
Purity: 97%
CAS No.: 83-07-8
Molecular Formula: C11H13N3O
Formula Weight: 203.25
Physical Form: Powder
Appearance: Yellow to pale brown
Melting Point: 106-110°
Beilstein Reference: 181635
Merck Reference: 14,591
MDL No.: MFCD00003145
Formula:
C₁₁H₁₃N₃O MW: 203.24 g/mol Melting Pt: 106…110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003145 CAS Number: 83-07-8 EINECS: 201-452-3 Merck Index: 14,591 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...