Dansyl chloride 96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Used for N terminal amino acid and peptide labeling
Purity: 96+%
CAS No.: 605-65-2
Molecular Formula: C12H12ClNO2S
Formula Weight: 269.75
Storage Temperature: 2°C to 8°C
Odor: Odorless
Beilstein Reference: 2217205
Merck Reference: 14,2814
UN No.: 3261
MDL No.: MFCD00003985
Formula:
C₁₂H₁₂ClNO₂S Melting Pt: 69…75°C Storage Temperature: Refrigerator |
MDL Number:
MFCD00003985 CAS Number: 605-65-2 EINECS: 210-092-6 UN: 3261 ADR: 8,III Merck Index: 14,2814 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...