Diethyl aminomalonate hydrochloride 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Diethyl 2-aminomalonate hydrochloride
A13681-18
A13681-30
A13681-09
AAA13681-18EA
118
USD
AAA13681-18
AAA13681-30
AAA13681-09
Diethyl aminomalonate hydrochloride 98%
Diethyl aminomalonate hydrochloride
Fieser: 15,132
Purity: 98%
CAS No.: 13433-00-6
Molecular Formula: C7H13NO4.HCl
Formula Weight: 211.65
Physical Form: Cryst powder
Sensitivity: Hygroscopic
Appearance: White to cream
Melting Point: ca 165° dec.
Beilstein Reference: 3568037
MDL No.: MFCD00012510
Formula:
C₇H₁₄ClNO₄ MW: 211.65 g/mol Melting Pt: ∼ 165 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00012510 CAS Number: 13433-00-6 EINECS: 236-556-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...