2-Iodophenol 98%

Supplier: Thermo Scientific Chemicals
Warning

A13599-22 A13599-14 A13599-06
AAA13599-22EA 365 USD
AAA13599-22 AAA13599-14 AAA13599-06
2-Iodophenol 98%
2-Iodophenol

Purity: 98% CAS No.: 533-58-4 Molecular Formula: C6H5IO Formula Weight: 220.01 Flash Point: 110°(230°F) Melting Point: 39-41° Boiling Point: 186-187°/160mm Beilstein Reference: 1855300 Merck Reference: 14,5035 MDL No.: MFCD00013963

Formula: C₆H₅IO
Boiling Pt: 186…187 °C/160 mm
Melting Pt: 39…41 °C
Density: 1.947
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00013963
CAS Number: 533-58-4
EINECS: 208-569-9
UN: 2811
ADR: 6.1,III
Merck Index: 14,5035

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR