(+)-Rutin trihydrate 95%

Supplier: Thermo Scientific Chemicals

A13570-22 A13570-14 A13570-36
AAA13570-22EA 93.7 USD
AAA13570-22 AAA13570-14 AAA13570-36
(+)-Rutin trihydrate 95%
(+)-Rutin trihydrate

Purity: 97% CAS No.: 250249-75-3 Molecular Formula: C27H30O16.3H2O Formula Weight: 664.58 (610.52anhy) Melting Point: 184° dec. Beilstein Reference: 75455 Optical Rotation: +9° (c=0.5 in ethanol) MDL No.: MFCD00149490

Formula: C₂₇H₃₀O₁₆·3H₂O
Melting Pt: 184 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00149490
CAS Number: 250249-75-3
EINECS: 205-814-1
Merck Index: 14,3802

Order Now

Specification Test Results

Optical rotation +9° (c=0.5 in ethanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR