1-Cetylpyridinium chloride monohydrate 98%

Supplier: Thermo Scientific Chemicals
Danger

A13499-14 A13499-22 A13499-36
AAAA13499-14EA 22.81 USD
AAAA13499-14 AAA13499-22 AAA13499-36
1-Cetylpyridinium chloride monohydrate 98%
1-Cetylpyridinium chloride monohydrate

MDL: MFCD00149977 Beilstein: 3578606 Soluble in water, alcohol, chloroform.


Purity: 98% CAS No.: 6004-24-6 Molecular Formula: C21H38ClN.H2O Formula Weight: 358.02 (339.99anhy) Melting Point: 83-86° Solubility: Soluble in water, alcohol, chloroform Beilstein Reference: 3578606 Merck Reference: 14,2032 UN No.: 2811 MDL No.: MFCD00149977

Formula: C₂₁H₃₈ClN·1H₂O
Melting Pt: 83…86 °C
Storage Temperature: Ambient
MDL Number: MFCD00149977
CAS Number: 6004-24-6
EINECS: 204-593-9
UN: 2811
ADR: 6.1,III
Merck Index: 14,2032

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR