2-Nitrotoluene 99+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A13446-22
A13446-36
A13446-0E
AAA13446-22EA
30.9
USD
AAA13446-22
AAA13446-36
AAA13446-0E
2-Nitrotoluene 99+%
2-Nitrotoluene
Purity: 99+%
CAS No.: 88-72-2
Molecular Formula: C7H7NO2
Formula Weight: 137.14
Flash Point: 106°(222°F)
Refractive Index: 1.5450
Melting Point: -4°
Boiling Point: 224-226°
Beilstein Reference: 1907580
Merck Reference: 14,6650
UN No.: 1664
MDL No.: MFCD00007157
Formula:
C₇H₇NO₂ MW: 137.14 g/mol Boiling Pt: 221.7 Melting Pt: -1 Density: 1.166 Flash Pt: 106 °C (222 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007157 CAS Number: 88-72-2 EINECS: 201-853-3 UN: 1664 ADR: 6.1 II Merck Index: 14,6650 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...