Lithium hydroxide, anhydrous 98%

Supplier: Thermo Scientific Chemicals
Danger

013407-22 013407-14 013407-36
AA13407-22EA 85.55 USD
AA13407-22 AA13407-14 AAA13407-36
Lithium hydroxide, anhydrous 98%
Lithium hydroxide

Anhydrous


Purity: 98% CAS No.: 1310-65-2 Molecular Formula: LiOH Formula Weight: 23.95 Physical Form: -4+14 Mesh Density: 2.54 Appearance: White Odor: Odorless pH: 14 Melting Point: 462° Boiling Point: 924° Solubility: Soluble in water. Slightly soluble in alcohol Merck Reference: 14,5534 UN No.: 2680 MDL No.: MFCD00011095

Formula: LiOH
Boiling Pt: 924 °C
Melting Pt: 462 °C
Density: 2.54
Storage Temperature: Ambient
MDL Number: MFCD00011095
CAS Number: 1310-65-2
EINECS: 215-183-4
UN: 2680
ADR: 8,II
Merck Index: 14,5534

Order Now

Specification Test Results

Form -4+14 Mesh


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR