1,2-Epoxycyclohexane 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Epoxycyclohexane, Cyclohexene oxide
A13185-0E
A13185-22
A13185-36
AAA13185-0EEA
614
USD
AAA13185-0E
AAA13185-22
AAA13185-36
1,2-Epoxycyclohexane 98+%
1,2-Epoxycyclohexane
MDL: MFCD00005162
Beilstein: 383568
Insoluble in water
Purity: 98+%
CAS No.: 286-20-4
Molecular Formula: C6H10O
Formula Weight: 98.15
Flash Point: 24°(75°F)
Refractive Index: 1.4520
Melting Point: -40°
Boiling Point: 129-130°
Solubility: Insoluble in water
Beilstein Reference: 383568
UN No.: 2920
MDL No.: MFCD00005162
Formula:
C₆H₁₀O MW: 98.14 g/mol Boiling Pt: 129 Melting Pt: -27 Density: 0.972 Flash Pt: 24 °C (75 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005162 CAS Number: 286-20-4 EINECS: 206-007-7 UN: 2920 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...