Dibenzo-18-crown-6-(dibenzo-1,4,7,10,13,16-hexaoxacyclooctadecane, tin(II) ionophore I) 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Dibenzo-18-crown-6
A13133-14
A13133-06
AAA13133-14EA
90.53
USD
AAA13133-14
AAA13133-06
Dibenzo-18-crown-6-(dibenzo-1,4,7,10,13,16-hexaoxacyclooctadecane, tin(II) ionophore I) 98+%
Dibenzo-18-crown-6 (Dibenzo-1,4,7,10,13,16-hexaoxacyclooctadecane, tin (II) ionophore I)
Fieser: 6,159 8,128 9,126
Purity: 98+%
CAS No.: 14187-32-7
Molecular Formula: C20H24O6
Formula Weight: 360.41
Melting Point: 160-164°
Boiling Point: 380-384°/679mm
Merck Reference: 14,2602
MDL No.: MFCD00005098
Formula:
C₂₀H₂₄O₆ Boiling Pt: 380…384 °C/679 mm Melting Pt: 160…164 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005098 CAS Number: 14187-32-7 EINECS: 238-041-3 Merck Index: 14,2602 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...