tert-Butylchlorodimethylsilane 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
tert-Butyldimethylsilyl Chloride, Chloro-tert-butyldimethylsilane, t-Butyldimethylchlorosilane, tert-Butyldimethylchlorosilane, tert-Butyldimethylsilyl chloride, TBDMS chloride
A13064-06
A13064-22
A13064-14
AAA13064-06EA
32.5
USD
AAA13064-06
AAA13064-22
AAA13064-14
tert-Butylchlorodimethylsilane 97%
tert-Butylchlorodimethylsilane
Purity: 97%
CAS No.: 18162-48-6
Molecular Formula: C6H15ClSi
Formula Weight: 150.73
Flash Point: 22°(72°F)
Odor: Pungent
Boiling Point: 124-126°
Beilstein Reference: 505999
UN No.: 2925
MDL No.: MFCD00000501
Formula:
C₆H₁₅ClSi Boiling Pt: 124…126 °C Melting Pt: 86…89 °C Density: 0.81 Flash Pt: 22 °C (72 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000501 CAS Number: 18162-48-6 EINECS: 242-042-4 UN: 2921 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...