Pyridine-2-carbonitrile 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Purity: 99%
CAS No.: 100-70-9
Molecular Formula: C6H4N2
Formula Weight: 104.11
Flash Point: 89°(192°F)
Refractive Index: 1.5290
Odor: Characteristic
pH: 8.4
Melting Point: 26-28°
Boiling Point: 212-214°
Beilstein Reference: 107710
MDL No.: MFCD00006218
Formula:
C₆H₄N₂ Boiling Pt: 212…214 °C Melting Pt: 26…28 °C Density: 1.081 Flash Pt: 89 °C (192 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006218 CAS Number: 100-70-9 EINECS: 202-880-3 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...