Dysprosium ≥99.9% (REO, rare earth oxide basis), powder -40 mesh

Supplier: Thermo Scientific Chemicals
Danger

012897-14 012897-06
AA12897-14EA 555 USD
AA12897-14 AAA12897-06
Dysprosium ≥99.9% (REO, rare earth oxide basis), powder -40 mesh
Dysprosium

MDL: MFCD00010982


Purity: 99.9% CAS No.: 7429-91-6 Physical Form: Packaged under argon Sensitivity: Air and Moisture Sensitive Appearance: Silver Odor: Odorless UN No.: 3089 MDL No.: MFCD00010982

Formula: Dy
Boiling Pt: 2335 °C (1013 hPa)
Melting Pt: 1409 °C
Density: 8.55 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010982
CAS Number: 7429-91-6
EINECS: 231-073-9
UN: 3089
ADR: 4.1,II
Merck Index: 12,03531

Order Now

Specification Test Results

Form Packaged under argon


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR