1-Heptanol 99%

Supplier: Thermo Scientific Chemicals
Warning

A12793-AP A12793-AE A12793-0F
AAA12793-APEA 40.4 USD
AAA12793-AP AAA12793-AE AAA12793-0F
1-Heptanol 99%
1-Heptanol

Purity: 99% CAS No.: 111-70-6 Molecular Formula: C7H16O Formula Weight: 116.20 Flash Point: 73°(163°F) Physical Form: Liquid Density: 0.820 Refractive Index: 1.4240 Appearance: Colorless Odor: Characteristic Melting Point: -34° Boiling Point: 175-176° Beilstein Reference: 1731686 Merck Reference: 14,4661 MDL No.: MFCD00002986

Formula: H₃C(CH₂)₅CH₂OH
Boiling Pt: 175…176 °C
Melting Pt: –34 °C
Density: 0.820
Flash Pt: 73 °C(163 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002986
CAS Number: 111-70-6
EINECS: 203-897-9
Merck Index: 14,4661

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR