Pentafluoropropionic acid 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Perfluoropropionic acid, Pentafluoropropanoic acid, Perfluoropropanoic acid, 2,2,3,3,3-Pentafluoropropionic acid
A12791-14
A12791-22
A12791-36
AAA12791-14EA
121.51
USD
AAA12791-14
AAA12791-22
AAA12791-36
Pentafluoropropionic acid 97%
Pentafluoropropionic acid
Purity: 97%
CAS No.: 422-64-0
Molecular Formula: CF3CF2CO2H
Formula Weight: 164.03
Refractive Index: 1.2840
Odor: Rancid
Boiling Point: 95-97°
Beilstein Reference: 1773387
UN No.: 3265
MDL No.: MFCD00004170
Formula:
C₃HF₅O₂ Boiling Pt: 95…97 °C Density: 1.576 Storage Temperature: Ambient |
MDL Number:
MFCD00004170 CAS Number: 422-64-0 EINECS: 207-021-6 UN: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...