Pyromellitic dianhydride 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A12712-22
A12712-0E
A12712-36
AAA12712-22EA
53.9
USD
AAA12712-22
AAA12712-0E
AAA12712-36
Pyromellitic dianhydride 97%
Pyromellitic dianhydride
Purity: 97%
CAS No.: 89-32-7
Molecular Formula: C10H2O6
Formula Weight: 218.12
Flash Point: 380°(716°F)
Melting Point: 284-288°
Boiling Point: 305-310°/30mm
Beilstein Reference: 213583
MDL No.: MFCD00005005
Formula:
C₁₀H₂O₆ Boiling Pt: 305…310 °C/30 mm Melting Pt: 284…288 °C Density: 1.68 Flash Pt: 380 °C(716 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005005 CAS Number: 89-32-7 EINECS: 201-898-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...