Nicotinic acid (Niacin) 99%

Supplier: Thermo Scientific Chemicals
Warning

A12683-0B A12683-30 A12683-0I
AAA12683-0BEA 101.43 USD
AAA12683-0B AAA12683-30 AAA12683-0I
Nicotinic acid (Niacin) 99%
Nicotinic acid (Niacin)

Vitamin of the B complex with hypolipidemic properties occurring in various animal and plant tissues. Required by the body for the formation of coenzymes NAD and NADP


Purity: 99% CAS No.: 59-67-6 Molecular Formula: C6H5NO2 Formula Weight: 123.11 Flash Point: 293°(559°F) Physical Form: Powder Density: 1.4 Appearance: White Odor: Odorless pH: 2.7 Melting Point: 236-239° Beilstein Reference: 109591 Merck Reference: 14,6525 MDL No.: MFCD00006391


Vitamin of the B complex with hypolipidemic properties occurring in various animal and plant tissues. Required by the body for the formation of coenzymes NAD and NADP

Formula: C₆H₅NO₂
Boiling Pt: 258 °C (1013 hPa)
Melting Pt: 236…239 °C
Density: 1.4
Flash Pt: 293 °C(559 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006391
CAS Number: 59-67-6
EINECS: 200-441-0
Merck Index: 14,6525

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR