Indole-3-carboxylic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1H-Indole-3-carboxylic acid
A12634-14
A12634-03
A12634-06
AAA12634-14EA
356
USD
AAA12634-14
AAA12634-03
AAA12634-06
Indole-3-carboxylic acid 98%
Indole-3-carboxylic acid
Purity: 98%
CAS No.: 771-50-6
Molecular Formula: C9H7NO2
Formula Weight: 161.16
Melting Point: ca 239° dec.
Beilstein Reference: 129435
Formula:
C₉H₇NO₂ Melting Pt: ∼ 239 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00005624 CAS Number: 771-50-6 EINECS: 212-231-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...