Methylphosphonic acid 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Methanephosphonic acid
A12619-03
A12619-14
A12619-06
A12619-22
AAAA12619-03EA
32
USD
AAAA12619-03
AAA12619-14
AAA12619-06
AAA12619-22
Methylphosphonic acid 98%
Methylphosphonic acid
Purity: 98%
CAS No.: 993-13-5
Molecular Formula: CH5O3P
Formula Weight: 96.03
Melting Point: 103-107°
Beilstein Reference: 1739372
UN No.: 3261
MDL No.: MFCD00002137
Formula:
CH₅O₃P Melting Pt: 103…107 °C Density: 1.639 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002137 CAS Number: 993-13-5 EINECS: 213-607-2 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...