Pyrrole 98+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1H-Pyrrole

A12616-30 A12616-18 A12616-0B
AAA12616-30EA 156.41 USD
AAA12616-30 AAA12616-18 AAA12616-0B
Pyrrole 98+%
Pyrrole

Purity: 98+% CAS No.: 109-97-7 Molecular Formula: C4H5N Formula Weight: 67.09 Storage Temperature: 2°C to 8°C Flash Point: 38°(100°F) Refractive Index: 1.5090 Odor: Mild Boiling Point: 131° Beilstein Reference: 1159 Merck Reference: 14,8014 UN No.: 1992 MDL No.: MFCD00005216

Formula: C₄H₅N
MW: 67.09 g/mol
Boiling Pt: 131 °C
Melting Pt: -24
Density: 0.967
Flash Pt: 38 °C (100 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00005216
CAS Number: 109-97-7
EINECS: 203-724-7
UN: 1992
ADR: 3 III
Merck Index: 14,8014

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR