N,N,N',N'-Tetramethylethylenediamine (TEMED) 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Purity: 99%
CAS No.: 110-18-9
Molecular Formula: C6H16N2
Formula Weight: 116.21
Flash Point: 18°(62°F)
Physical Form: Liquid
Density: 0.775
Refractive Index: 1.4180
Sensitivity: Air and Moisture Sensitive
Appearance: Colorless to pale yellow
Odor: Unpleasant
Melting Point: -55°
Boiling Point: 120-122°
Beilstein Reference: 1732991
UN No.: 2372
MDL No.: MFCD00008335
Formula:
C₆H₁₆N₂ Boiling Pt: 120…122 °C Melting Pt: –55 °C Density: 0.775 Flash Pt: 18 °C (62 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008335 CAS Number: 110-18-9 EINECS: 203-744-6 UN: 2372 ADR: 3,II |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...