Piperidine 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Fieser: 1,886 2,332 4,393 6,472 7,293 19,272
Purity: 99%
CAS No.: 110-89-4
Molecular Formula: C5H11N
Formula Weight: 85.15
Flash Point: 3°(37°F)
Physical Form: Liquid
Density: 0.862
Refractive Index: 1.4520
Sensitivity: Air Sensitive
Appearance: Colorless
Melting Point: -11°
Boiling Point: 105-106°
Beilstein Reference: 102438
Merck Reference: 14,7468
UN No.: 2401
MDL No.: MFCD00005979
Formula:
C₅H₁₁N Boiling Pt: 105…106 °C Melting Pt: –11 °C Density: 0.862 Flash Pt: 3 °C (37 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005979 CAS Number: 110-89-4 EINECS: 203-813-0 UN: 2401 ADR: 8,I Merck Index: 14,7468 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...