4-Aminopyridine 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A12405-30
A12405-0B
A12405-18
AAA12405-30EA
270
USD
AAA12405-30
AAA12405-0B
AAA12405-18
4-Aminopyridine 98%
4-Aminopyridine
Purity: 98%
CAS No.: 504-24-5
Molecular Formula: C5H6N2
Formula Weight: 94.12
Flash Point: 164°(327°F)
Odor: Unpleasant
Boiling Point: 273-274°
Beilstein Reference: 105782
Merck Reference: 14,3933
UN No.: 2671
MDL No.: MFCD00006439
Formula:
C₅H₆N₂ Boiling Pt: 273…274 °C Melting Pt: 157…161 °C Density: 1.26 Flash Pt: 164 °C (327 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006439 CAS Number: 504-24-5 EINECS: 207-987-9 UN: 2671 ADR: 6.1,II Merck Index: 14,3933 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...