Benzo[b]thiophene 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Thianaphthene, 1-Benzothiophene

A12328-22 A12328-14 A12328-06
AAA12328-22EA 281.48 USD
AAA12328-22 AAA12328-14 AAA12328-06
Benzo[b]thiophene 98+%
Benzo[b]thiophene

Purity: 98+% CAS No.: 95-15-8 Molecular Formula: C8H6S Formula Weight: 134.20 Flash Point: 91°(195°F) Odor: Unpleasant Boiling Point: 221-222° Beilstein Reference: 80580 Merck Reference: 14,9303 UN No.: 3077 MDL No.: MFCD00005864

Formula: C₈H₆S
MW: 134.2 g/mol
Boiling Pt: 221…222 °C
Melting Pt: 28…32 °C
Density: 1.149
Flash Pt: 91 °C (195 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005864
CAS Number: 95-15-8
EINECS: 202-395-7
UN: 3077
ADR: 9 III
Merck Index: 14,9303

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR