4-Bromo-3-methylanisole 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-Bromo-4-methoxy-2-methylbenzene, 2-Bromo-5-methoxytoluene
A12318-18
A12318-09
A12318-30
AAA12318-18EA
204
USD
AAA12318-18
AAA12318-09
AAA12318-30
4-Bromo-3-methylanisole 97%
4-Bromo-3-methylanisole
Purity: 97%
CAS No.: 27060-75-9
Molecular Formula: C8H9BrO
Formula Weight: 201.06
Flash Point:
Formula:
C₈H₉BrO Boiling Pt: 111…112 °C (15 mmHg) Density: 1.450 Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00060687 CAS Number: 27060-75-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...