Stearic acid 98%
Supplier: Thermo Scientific Chemicals
A saturated fatty acid
Purity: 98%
CAS No.: 57-11-4
Molecular Formula: CH3(CH2)16CO2H
Formula Weight: 284.48
Flash Point: 196°(384°F)
Physical Form: Flakes or powder
Density: 0.941
Appearance: White
Odor: Odorless
Melting Point: 69-72°
Boiling Point: 232°/15mm
Beilstein Reference: 608585
Merck Reference: 14,8804
MDL No.: MFCD00002752
A saturated fatty acid
Formula:
C₁₈H₃₆O₂ Boiling Pt: 232 °C/15 mm Melting Pt: 69…72 °C Density: 0.941 Flash Pt: 196 °C(384 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002752 CAS Number: 57-11-4 EINECS: 200-313-4 Merck Index: 14,8804 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...