Rubidium carbonate 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12240-06
A12240-14
A12240-22
AAA12240-06EA
37.36
USD
AAA12240-06
AAA12240-14
AAA12240-22
Rubidium carbonate 99%
Rubidium carbonate
CAS: 584-09-8
MDL: MFCD00011188
Formula:
CO₃Rb₂ Melting Pt: 837 °C Density: 2,43 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011188 CAS Number: 584-09-8 EINECS: 209-530-9 |
Specification Test Results
Assay Minimum | 99 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...