4'-Bromoacetophenone 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
p-Bromoacetophenone, 1-(4-Bromophenyl)ethanone
A12222-14
A12222-22
A12222-36
AAA12222-14EA
43
USD
AAA12222-14
AAA12222-22
AAA12222-36
4'-Bromoacetophenone 98%
4'-Bromoacetophenone
MDL: MFCD00000105
Beilstein Registry No.: 386015
Purity: 98%
CAS No.: 99-90-1
Molecular Formula: C8H7BrO
Formula Weight: 199.05
Flash Point:
Formula:
C₈H₇BrO Boiling Pt: 255…256 °C Melting Pt: 49…53 °C Density: 1.736 g/cm³ (20 °C) Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000105 CAS Number: 99-90-1 EINECS: 202-799-3 UN: 2811 ADR: 6.1,II Merck Index: 14,1403 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...