Sulfur trioxide pyridine complex 98% active SO₃ ∼ 48-50%

Supplier: Thermo Scientific Chemicals
Warning

A12202-18 A12202-0B A12202-30 A12202-09
AAA12202-18EA 64.31 USD
AAA12202-18 AAA12202-0B AAA12202-30 AAAA12202-09
Sulfur trioxide pyridine complex 98% active SO₃ ∼ 48-50%
Sulphur trioxide pyridine complex

Fieser: 1,1127 2,393 3,275 4,473


Purity: 98% CAS No.: 26412-87-3 Molecular Formula: C5H5NO3S Formula Weight: 159.16 Melting Point: ca 155-165° Beilstein Reference: 3704116 UN No.: 3261 MDL No.: MFCD00012437

Formula: C₅H₅NO₃S
Melting Pt: ∼ 155…165 °C
Storage Temperature: Ambient
MDL Number: MFCD00012437
CAS Number: 26412-87-3
EINECS: 247-683-3
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR