Indene 90+% stabilised
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1H-Indene
Stabilization:
Stabilised with 4-tert-butylpyrocatechol (TBC) 0.01%
A12192-AD
A12192-AK
A12192-AU
AAA12192-ADEA
31.6
USD
AAA12192-AD
AAA12192-AK
AAA12192-AU
Indene 90+% stabilised
Indene
MDL: MFCD00003777
Beilstein Registry No.: 635873
Purity: 90+%
CAS No.: 95-13-6
Molecular Formula: C9H8
Formula Weight: 116.16
Storage Temperature: 2°C to 8°C
Flash Point: 58°(136°F)
Refractive Index: 1.5700
Odor: Aromatic
Melting Point: ca -2°
Boiling Point: 181-182°
Beilstein Reference: 635873
Merck Reference: 14,4939
UN No.: 3295
MDL No.: MFCD00003777
Formula:
C₉H₈ Boiling Pt: 181…182 °C Melting Pt: ∼ 2 °C Density: 0.996 Flash Pt: 58 °C (136 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00003777 CAS Number: 95-13-6 EINECS: 202-393-6 UN: 3295 ADR: 3,III Merck Index: 14,4939 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...