Potassium cyanide ≥96.0% ACS

Supplier: Thermo Scientific Chemicals
Danger

012136-18 012136-30 012136-A1
AAA12136-18EA 90.45 USD
AAA12136-18 AAA12136-30 AAA12136-A1
Potassium cyanide ≥96.0% ACS
Potassium cyanide
Formula: KCN
MW: 65.12 g/mol
Boiling Pt: 1625 °C
Melting Pt: 634 °C
Density: 1.52
Storage Temperature: Ambient
MDL Number: MFCD00011397
CAS Number: 151-50-8
EINECS: 205-792-3
UN: 1680
ADR: 6.1,I
Merck Index: 14,7626

Order Now

Specification Test Results

Form Granular

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR