Dimethylammonium chloride 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Dimethylamine hydrochloride
A12133-0B
A12133-0I
A12133-30
AAA12133-0BEA
89.9
USD
AAA12133-0B
AAA12133-0I
AAA12133-30
Dimethylammonium chloride 98+%
Dimethylammonium chloride
MDL: MFCD00012477
Beilstein Registry No.: 3589311
Purity: 98+%
CAS No.: 506-59-2
Molecular Formula: (CH3)2NH.HCl
Formula Weight: 81.55
Melting Point: 168-174°
Beilstein Reference: 3589311
Merck Reference: 14,3228
MDL No.: MFCD00012477
Formula:
C₂H₈ClN Melting Pt: 168…174 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012477 CAS Number: 506-59-2 EINECS: 208-046-5 Merck Index: 14,3228 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...