Tris(2-methylphenyl)phosphine 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Tris(o-tolyl)phosphine, Tri-o-tolylphosphine

A12093-03 A12093-06 A12093-14
AAA12093-03EA 27.7 USD
AAA12093-03 AAA12093-06 AAA12093-14
Tris(2-methylphenyl)phosphine 98+%
Tris(2-methylphenyl)phosphine

MDL: MFCD00008514 Beilstein Registry No.: 661212


Purity: 98+% CAS No.: 6163-58-2 Molecular Formula: C21H21P Formula Weight: 304.37 Melting Point: 122-127° Beilstein Reference: 661212

Formula: C₂₁H₂₁P
Boiling Pt: 187 - 192 °C (0.05 -rr)
Melting Pt: 122…127 °C
Storage Temperature: Ambient
MDL Number: MFCD00008514
CAS Number: 6163-58-2
EINECS: 228-193-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR