2-Phenylbenzoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Biphenyl-2-carboxylic acid, 2-Biphenylcarboxylic acid, [1,1'-Biphenyl]-2-carboxylic acid

A12049-14 A12049-06 A12049-22
AAA12049-14EA 78.7 USD
AAA12049-14 AAA12049-06 AAAA12049-22
2-Phenylbenzoic acid 98%
2-Phenylbenzoic acid

MDL: MFCD00002463 Beilstein Registry No.: 974075


Purity: 98% CAS No.: 947-84-2 Molecular Formula: C13H10O2 Formula Weight: 198.22 Odor: Odorless Melting Point: 110-114° Boiling Point: 343-344° Beilstein Reference: 974075 MDL No.: MFCD00002463

Formula: C₁₃H₁₀O₂
Boiling Pt: 343…344 °C
Melting Pt: 110…114 °C
Density: 1.458 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002463
CAS Number: 947-84-2
EINECS: 213-432-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR