Guanine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Amino-1H-purin-6(7H)-one

A12024-14 A12024-22 A12024-36
AAA12024-14EA 49 USD
AAA12024-14 AAA12024-22 AAAA12024-36
Guanine 98%
Guanine

MDL: MFCD00071533 Beilstein Registry No.: 9680


Purity: 98% CAS No.: 73-40-5 Molecular Formula: C5H5N5O Formula Weight: 151.13 Storage Temperature: 2°C to 8°C Melting Point: 300° Beilstein Reference: 9680 Merck Reference: 14,4564 MDL No.: MFCD00071533

Formula: C₅H₅N₅O
Melting Pt: >300 °C
Density: 1.67 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00071533
CAS Number: 73-40-5
EINECS: 200-799-8
Merck Index: 14,4564

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR