2-Deoxy-D(-)-ribose 99%

Supplier: Thermo Scientific Chemicals

A11990-03 A11990-06 A11990-14
AAA11990-03EA 26.2 USD
AAA11990-03 AAA11990-06 AAA11990-14
2-Deoxy-D(-)-ribose 99%
2-Deoxy-D(-)-ribose
Formula: C₅H₁₀O₄
Boiling Pt: 292 °C (1013 hPa)
Melting Pt: 76…85 °C
Storage Temperature: Ambient
MDL Number: MFCD00135904
CAS Number: 533-67-5
EINECS: 208-573-0
Merck Index: 14,2908

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR