5-Bromoanthranilic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Amino-5-bromobenzoic acid

A11915-06 A11915-22 A11915-14
AAA11915-06EA 54 USD
AAA11915-06 AAA11915-22 AAA11915-14
5-Bromoanthranilic acid 98%
5-Bromoanthranilic acid

MDL: MFCD00007823 Beilstein Registry No.: 639028


Purity: 98% CAS No.: 5794-88-7 Molecular Formula: C7H6BrNO2 Formula Weight: 216.03 Melting Point: ca 215° dec. Beilstein Reference: 639028 Merck Reference: 14,1405 MDL No.: MFCD00007823

Formula: C₇H₆BrNO₂
Boiling Pt: 342 °C (1013 hPa)
Melting Pt: ∼ 215 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00007823
CAS Number: 5794-88-7
EINECS: 227-338-3
UN: 2811
ADR: 6.1,III
Merck Index: 14,1405

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR