4-Methyl-2-pentanone 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
MDL: MFCD00008938
Beilstein Registry No.: 605399
Purity: 99%
CAS No.: 108-10-1
Molecular Formula: C6H12O
Formula Weight: 100.16
Flash Point: 13°(55°F)
Refractive Index: 1.3960
Odor: Mild
Melting Point: -84°
Boiling Point: 117-118°
Beilstein Reference: 605399
Merck Reference: 14,5207
UN No.: 1245
MDL No.: MFCD00008938
Formula:
(CH₃)₂CHCH₂COCH₃ Boiling Pt: 117…118 °C Melting Pt: –84 °C Density: 0.801 Flash Pt: 13 °C (55 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008938 CAS Number: 108-10-1 EINECS: 203-550-1 UN: 1245 ADR: 3,II Merck Index: 14,5207 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...