Sodium-DL-3-hydroxybutyrate 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
Sodium 3-hydroxybutanoate, DL-3-Hydroxybutyric acid Sodium salt
A11613-06
A11613-14
AAA11613-06EA
38.99
USD
AAA11613-06
AAA11613-14
Sodium-DL-3-hydroxybutyrate 98%
Sodium-DL-3-hydroxybutyrate
Purity: 98%
CAS No.: 150-83-4
Molecular Formula: C4H7NaO3
Formula Weight: 126.09
Melting Point: 161-167°
Beilstein Reference: 4824866
MDL No.: MFCD00016716
Formula:
C₄H₈O₃ Melting Pt: 161…167 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016716 CAS Number: 150-83-4 EINECS: 205-774-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...